missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LTF/Lactoferrin ELISA Kit

Codice prodotto. 16672103
Click to view available options
Quantità:
96 Tests
Dimensione della confezione:
96 tests
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16672103

Marca: Invitrogen™ EH309RB

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

Human LTF/Lactoferrin quantitates human LTF/Lactoferrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LTF/Lactoferrin.

Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; Kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; Ltf; minisatellite 10 r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin
Intervallo saggio 1.96 to 480 ng/mL
Assay Sensitivity 1.96 ng/mL
Coniugato HRP
Product Type ELISA
Tipo di campione Plasma, Serum, Supernatant
Da utilizzare con (apparecchiatura) Colorimetric Microplate Reader
ID gene (immissione) 4057
Simbolo del gene LTF
Interassay CV <12%
Intraassay CV <10
Contenuto kit Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantità 96 Tests
Status giuridico RUO
Volume del campione Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Requisiti di stoccaggio 2°C to 8°C
Bersaglio Lactoferrin
Durata del test 1 hrs 20 mins
Tempo totale saggio 4 hrs 45 mins
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato