missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MMP-12 Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Marca: R&D Systems™ LMPM919
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plateSpecifica
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
27.60 to 6700 pg/mL | |
MMP12 | |
Cell Culture Supernates (10μL),Serum (10μL),Heparin Plasma (10μL),Platelet-poor Heparin Plasma (10μL),Saliva (10 ug),Urine (10μL) | |
|
1 Kit | |
MMP-12 | |
0.7 pg/mL | |
4321 | |
Cell Culture Supernatants, Serum, Plasma (Heparin), Platelet-poor Plasma (Heparin), Saliva, Urine | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto