missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 26 publications
Marca: R&D Systems MAB1455
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC.
Specifica
| Albumin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2-1 ug/mL, Simple Western 1-10 ug/mL, Immunohistochemistry 0.1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 0.05-25 ug/mL | |
| ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 213 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, In Situ Hybridization (ISH) | |
| 188835 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALB | |
| Human Serum Albumin | |
| 100 μg | |
| Primary | |
| Detects human Serum Albumin in direct ELISAs and Western blots. Does not detect mouse Serum Albumin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto