missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Troponin I ProQuantum Immunoassay Kit
The ProQuantum Human Troponin I Immunoassay Kit is designed to provide quantitative measurements of human Troponin I in small sample volumes.
Marca: Invitrogen™ A46074
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifica
cardiac troponin I; cardiac troponin I isoform; cardiomyopathy, dilated 2 A (autosomal recessive); CMD1FF; CMD2A; CMH7; cTNI; MGC116817; RCM1; Tn1; TnI; TNNC1; Tnni3; Tpn1; troponin 1, type 3; troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I3, cardiac type | |
<0.3 pg/mL | |
Plasma, Serum, Supernatant | |
P19429 | |
Assay Dilution Buffer Antibody-Conjugate A Antibody-Conjugate B Antibody-Conjugate Dilution Buffer Standard Master Mix Ligase | |
Research Use Only | |
-20°C | |
30 min |
0.32 to 25,000 pg/mL | |
ELISA | |
qPCR Instruments | |
TNNI3 | |
96 Tests | |
Serum, 2-5 μL; Plasma, 2-5 μL; Supernatant, 2-5 μL | |
Cardiac Troponin I | |
2 hr |