missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Troponin I/TNNI3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB68871
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Troponin I Monoclonal specifically detects Troponin I in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifica
| Troponin I | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1, TNNC1troponin I, cardiac, TNNI3,, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7137 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 679021 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TNNI3 | |
| Human cardiac Troponin I purified from human heart | |
| 100 μg | |
| Primary | |
| Detects human Cardiac Troponin I in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido