missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PeproTech® Human VISTA Fc Recombinant Protein
Recombinant Protein
Marca: PeproTech® 310-45-500UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Recombinant Human VISTA Fc is a 88.2 kDa homodimeric protein of 395 amino acids consisting of the extracellular domain of the mature sequence, as well as a 230-residue C-terminal Fc region linked by two glycines. As a result of glycosylation, the Recombinant Human VISTA Fc migrates to an apparent molecular weight of approximately 60-70 kDa by SDS-PAGE gel, under reducing conditions.
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Specifica
Q9H7M9 | |
Lyophilized | |
44.2 | |
-20°C | |
<1 | |
Determined by its ability to bind immobilized human VSIG3 Fc in a functional ELISA | |
Recombinant | |
CHO cells |
Functional Assay | |
protein with no preservative | |
500 μg | |
RUO | |
VISTA | |
Unconjugated | |
Fc-tag | |
purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto