Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Codice prodotto. 10553764
Click to view available options
Quantità:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
Con additivi:
Calcium, Magnesium
Calcium, Magnesium, Phenol Red
Phenol Red
Phenol Red, Calcium, Magnesium
Senza additivi:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
Dimensione della confezione:
1000mL
100mL
500mL
pezzi 6
Codice prodotto. 10553764
Marca: Cytiva SH30031.FS
Buffer cell culture growth with 1X balanced salt solutions
  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentrazione 1 X
Forma Liquid
Product Type Hanks' Balanced Salt Solution
Sterilization Method 0.1 μm Sterile-filtered
Con additivi Phenol Red
Senza additivi No Calcium, No Magnesium
Livello di endotossine =1.0EU/mL
pH 7.0 to 7.4
Quantità 6 x 500 mL