Learn More
Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA), average age at time of collection is 6 to 8 years
Marca: Cytiva SH30528.05
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Specifica
6 to 8 years | |
Adult Bovine Serum | |
Triple-filtered | |
HyClone™ |
New Zealand | |
3000 mL | |
Irradiated | |
Tested According to European Pharmacopoeia Protocols |