missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyurea, 98%

Codice prodotto. 10502164 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
5 g
25 g
100 g
Imballaggio:
Bottiglia in vetro
Dimensione della confezione:
100g
25g
5g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10502164

Marca: Thermo Scientific Acros 151681000

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Hydroxyurea, 98%, CAS # 127-07-1, is a compound with antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities. It shows inhibitory activity against nucleic acid synthesis. | CAS: 127-07-1 | CH4N2O2 | 76.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hydroxyurea is a member of the class of urea that shows antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities.

• It has inhibitory activity against nucleic acid synthesis.

• It also inhibits ribonucleoside diphosphate reductase, an enzyme necessary to convert ribonucleoside diphosphate into deoxyribonucleoside diphosphate, preventing cells from leaving the G1/S phase of the cell cycle

Applications

• Hydroxyurea has an anti-retroviral activity, inhibiting human immunodeficiency virus type 1 (HIV-1) DNA synthesis in peripheral blood lymphocytes, decreasing the amount of intracellular deoxynucleotides

• This compound also has a radio sensitizing activity, maintaining cells in the radiation-sensitive G1 cell cycle phase and, consequently, interfering with DNA repair

• It has been used for cell cycle synchronization in human breast carcinoma cells

• This compound presents inhibitory activity in cell adhesion and motility assay in ccRCC Caki (HTB-47) and CD9 knockdown cells

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 127-07-1
Formula molecolare CH4N2O2
Molecular Weight (g/mol) 76.04
Numero MDL MFCD00007943
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Sinonimo hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifica

Materiale o nome chimico Hydroxyurea
Punto di fusione 142°C to 146°C
Colore White
Perdita all'essiccatura 1% max. (1 g, 60°C) (< 0.7 kPa)
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97.5% min. (on dried substance) (Iodometry)
Imballaggio Bottiglia in vetro
Formula lineare HONHCONH2
Quantità 100 g
Beilstein 03, 95
Indice di Merck 15, 4888
Informazioni di solubilità Solubility in water: freely soluble. Other solubilities: freely soluble in hot alcohol
Peso formulazione 76.04
Percent Purity 98%
Forma fisica Crystalline Powder
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Hydroxyurea
Avvertenza
  • Pericolo
Categoria di pericolo
  • Mutagenicità di cellule germinali Categoria 1B
  • Tossicità per la riproduzione Categoria 1A
Indicazioni di pericolo
  • H360D-Può nuocere al feto.
  • H340-Può provocare alterazioni genetiche.
Consigli di prudenza
  • P201-Procurarsi istruzioni specifiche prima dell'uso.
  • P308+P313-IN CASO di esposizione o di possibile esposizione, consultare un medico.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato