Learn More
Ansell™ HyFlex™ 11-616 Series Black and Green/Blue Polyurethane Light Weight Gloves
Click to view available options
Size:
6
7
8
9
10
11
Descrizione
- 18-gauge knitting technology allows this glove to be ultra-thin yet tough
- Ideal for workers who need a higher level of fit, touch and precision
- Ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries.
Specifica
Specifica
Materiale parte posteriore | Polyurethane |
Certificazione/Conformità | CE Mark, EN 388 : 3121 |
Materiale polso | Polyurethane |
Coating Material | Polyurethane |
Product Type | Lightweight Gloves |
Size | 8 |
Stile | 11-616 |
Stile polso guanto | Knit |
Rivestimento interno | Nylon |
Tipo di palmo | Coated-palm |