Learn More
Ansell™ HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
Click to view available options
Size:
6
7
8
9
10
11
Descrizione
- Patented Ansell Grip Technology™.
- Microscopic channels in the glove coating guide away the oil for secure glove-to-surface contact.
- Knitted construction
- Nylon liner
- Nitrile coating
- Palm-coated finish
- Knitwrist cuff
- Blue color
Specifica
Specifica
Materiale parte posteriore | Nitrile |
Certificazione/Conformità | ANSI, EN 388 : 3131 |
Materiale polso | Nitrile |
Coating Material | Nitrile |
Product Type | Mediumweight Gloves |
Size | 8 |
Stile | 11-920 |
Stile polso guanto | Knit |
Rivestimento interno | Nylon |
Tipo di palmo | Coated-palm |