Learn More
Thermo Scientific™ Hypersil GOLD™ PFP HPLC Columns
Codice prodotto. p-4523036 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
10 mm
20 mm
30 mm
50 mm
100 mm
150 mm
200 mm
250 mm
Diametro (sistema metrico):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
10 mm
21.2 mm
30 mm
50 mm
Dimensione particelle:
1.9 μm
3 μm
5 μm
Dimensione della confezione:
Pezzo
pezzi 3
pezzi 4
Codice prodotto. 10067364
Marca: Thermo Scientific™ 25405109070
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Use for analyzing difficult-to-resolve mixtures of halogenated compounds including pesticides.
Analyze difficult-to-resolve mixtures of halogenated compounds and non-halogenated polar compounds with Thermo Scientific™ Hypersil GOLD™ PFP (pentafluorophenyl) HPLC columns. These columns provide an alternative selectivity to C18 columns in reversed phase applications with excellent peak shape and sensitivity.
Unique selectivity
  • Increased retention and selectivity for positional isomers of halogenated compounds
  • Excellent selective analysis of non-halogenated compounds, in particular polar compounds containing hydroxyl, carboxyl, nitro, or other polar groups
  • Efficient separation of difficult mixtures containing closely related halogenated and polar compounds
The introduction of a fluorine group into the stationary phase causes significant changes in solute-stationary phase interaction and increases the selectivity and extra retention.
About the Hypersil GOLD family of HPLC columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3, and 5μm particle sizes for all your separation needs
TRUSTED_SUSTAINABILITY
Specifica
Formato colonna Preparative Column
Diametro (sistema metrico) 10 mm
Lunghezza (sistema metrico) 100 mm
Pressione max 5800 psi (400 bar)
Dimensione particelle 5 μm
pH 2 to 8
Dimensione del poro 175 Å
Surface Area 220 m²/g
Temperatura 60 °C
Tipo USP L43
Fase stazionaria PFP (Pentafluorophenylpropyl)
Product Line Hypersil GOLD
Materiale di impaccamento diossido di silicio ultrapuro completamente poroso, sferico
Carico di carbonio 8%
Tipo di colonna A fase inversa
Con tappo
Fase A fase inversa