missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Monoclonal Antibody (307426), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523587
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-27 Monoclonal antibody specifically detects IL-27 in Human samples. It is validated for Flow Cytometry.Specifica
IL-27 | |
Monoclonal | |
FITC | |
Ebi3 | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
Mouse | |
Protein A/G | |
RUO | |
10148, 246778 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
307426 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q14213, Q8NEV9 | |
Ebi3, IL27 | |
Mouse myeloma cell line NS0-derived recombinant human IL-27 Arg21-Lys229 of EBI-3 and Phe29-Pro243 of p28 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |