missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imipramine Monoclonal Antibody (IM12-342.3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA110723
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
MA1-10723 detects Imipramine. MA1-10723 has been successfully used in ELISA applications. The MA1-10723 immunogen is Imipramine, conjugated to KLH.
Imipramine Base is a synthetic tricyclic derivative, antidepressant Imipramine enhances monoamine neurotransmission in certain areas of the brain. It also induces sedation through histamine 1 receptor blockage; hypotension through beta-adrenergic blockage; and diverse parasympatholytic effects. Imipramine has less sedative effect than other members of its therapeutic family. It is used in major depression, dysthymia, bipolar depression, attention-deficit disorders, agoraphobia, and panic disorders. [National Center for Biotechnology Information. PubChem Compound Database; CID=3696].Specifica
Imipramine | |
Monoclonal | |
1.4 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG2b |
ELISA | |
IM12-342.3 | |
Unconjugated | |
Antideprin; C19H24N2; Deprenil; Deprimin; Deprinol; Depsonil; Dynaprin; Eupramin; Imipramil; Irmin; Janimine; Melipramin; Surplix; Tofranil | |
Imipramine, conjugated to KLH. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |