Learn More
Thermo Scientific Chemicals Inconel Cover for Crucible
Click to view available options
Capacità (sistema metrico):
10 mL
15 mL
20 mL
25 mL
45 mL
55 mL
75 mL
100 mL
250 mL
500 mL
Descrizione:
For 35886 crucible
For 35911 crucible
For 35932 crucible
For 35953 crucible
For 35974 crucible
For 36012 crucible
For 36016 crucible
For 36052 crucible
For 36056 crucible
For 36072 crucible
For 36090 crucible
For 36102 crucible
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Descrizione | For 35974 crucible |
| Materiale | Inconel |
| Capacità (sistema metrico) | 25 mL |
| Product Type | Inconel Crucible Cover |