missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H3N2 Virions Polyclonal Antibody, HRP
Goat Polyclonal Antibody
Marca: Invitrogen™ PA173048
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PA1-73048 antibody reacts with Influenza A virions H3N2. PA1-73048 has been successfully used in ELISA applications. The PA1-73048 antibody was raised against Influenza A, strain Texas 1/77 (H3N2).
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifica
| Influenza A H3N2 Virions | |
| Polyclonal | |
| HRP | |
| H3N2; H3N2 virions; Influenza H3N2 | |
| Influenza A, strain Texas 1/77 (H3N2). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1-2 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto