Learn More
Thermo Scientific™ LinerGOLD™ GC Liners
Codice prodotto. 11827290 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Diametro (sistema metrico) interno:
0.75 mm
1 mm
2 mm
3 mm
4 mm
5 mm
Mixed
Lunghezza (sistema metrico):
105 mm
105 mm
120 mm
120 mm
78.5 mm
Mixed
Quantità:
1
2
5
25
Product Type:
Direct Straight Liner
Double Taper Cyclo Liner
Mixed Liner Pack
PTV Baffle Liner (Siltek)
PTV Glass Straight Liner (Deactivated)
PTV Liner, Concentric Baffles
PTV Liner, Offset 5 Baffles
PTV Liner, Sintered Lining
PTV Liner, Tenax TA
PTV Liner, Three Baffles
PTV Silcosteel Liner for OC
PTV Siltek Metal Liner
PTV Siltek Metal Liner, Wool
PTV Split/Splitless Straight Liner
PTV Straight Liner
Precision Liner
Split FocusLiner
Split Straight Liner
Split/Splitless Liner
Split/Splitless Mini-Lam Liner
Split/Splitless Mixed Liner Sample Pack
Split/Splitless Straight Liner
Splitless FocusLiner
Splitless FocusLiner for 50 mm Needle
Splitless Liner
Splitless Liner, Double Taper
Splitless Liner, Single Taper
Splitless Liner, Single Taper, Wool
Splitless Precision Liner
Splitless Straight Liner
Splitless Straight Liner (Siltek)
Dimensione della confezione:
Pezzo
pezzi 2
pezzi 25
pezzi 5
Codice prodotto. 11827290
Marca: Thermo Scientific™ 45352054
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
  • Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
  • Lower detection limits when analyzing active and sensitive compounds
  • Excellent reproducibility gives confidence in your results from liner to liner
  • Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 5
Lunghezza (sistema metrico) 120 mm
Materiale di impaccamento None
Product Type PTV Straight Liner
Diametro (sistema metrico) interno 1 mm
Diametro (sistema metrico) esterno 2.75 mm
Da utilizzare con (apparecchiatura) Thermo Scientific Instruments