Learn More
Fisherbrand™ Ink Chart Recorders for Isotemp Ultra-Low Temperature Chest Freezers
Record seven days of temperature performance at temperatures to -150°C using Fisherbrand™ Isotemp Chart Recorders for Ultra-Low Temperature Freezers.
Marca: Fisherbrand™ 332166G12
1335.25 EUR valido fino al 2024-12-31
Usa il codice promo "21658" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Seven-day recorder
- Functions to -150°C
- Factory installed
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Factory Installed | |
-150°C | |
7 Days |
Chart Recorder | |
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |