missing translation for 'onlineSavingsMsg'
Learn More

WTW™ pH Benchtop Meter inoLab™ pH 7310

The inoLab™ pH 7310 with backlit graphic display is the right meter for precision measurements of pH and ORP potentials and the automatic documentation according to GLP/AQA in quality labs in all industries.

Marca:  WTW™ 1AA312

Dettagli aggiuntivi : Peso : 0.80000kg

Codice prodotto. 11791446

  • 1335.00€ / Pezzo
Spedizione stimata: 11-10-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

  • Precise pH meter with documentation function to use in quality assurance, pharmaceutical industry and everywhere GLP-supported documentation is required
  • Set with standard electrode with temperature sensor
  • Repeatable measuring results by automatic recognition of stable values
  • Monitoring of the optimum measuring range via CMC function
  • Secure and conveniant operation via text menus in a bright graphic display
  • Includes 3 in 1 pH electrode, meter with universal power supply, electrode arm and operation manual
  • pH electrode SenTix™ 41, buffers, 3mol/L KCl, software and USB cable
Specifica

Specifica

±0.005, ±0.01
±0.3, ±1
3 in 1 pH electrode, meter with universal power supply, electrode arm and operation manual. 3 in 1 pH electrode SenTix™ 41, buffers, 3mol/L KCl, software and USB cable
-2 to 20
±1200, ±0.3
1 to 3
CE, cETLUS
0.8 kg
Four 1.5V AA batteries or four 1.2V NiMH
Automatic/Manual
DIN or BNC/4mm banana
Up to 1000 hr. without backlight, 150 hr. with backlight
Up to 10 records calibration memory, 500 manual datasets/5000 automatic memory
±0.1K
For measurements/documentation according GLP/AQA, AC and battery operation
-2 to 20 ±0.01, -2 to 19.999 ±0.005
±(1200 ±1)mV
LCD, Graphic with Backlight
240 x 190 x 80 mm
3 Years
Yearly mean: <75%, 30 days/year: 95%, Other days: 85%
5°C to 55°C
inoLab™ Wastewater 7310 set 2 pH meter
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali