Learn More
Diversey™ IntelliCare Dispenser Hybrid
The IntelliCare Hybrid dispensing system can address all hand care needs with a versatile, modern, and durable platform that is compatible with both liquid/gels and foam products.
Marca: Diversey™ D7524179
111.59 EUR valido fino al 2024-12-31
Usa il codice promo "21676" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Dispenses 1300 mL bottles of hand soap and 1200 mL of alcohol based hand sanitizer
- Can dispense 0.4 mL or 1.00 mL per push
- Hybrid dispenser provides an automatic switch from touchless to manual if batteries died before replacement
- Near empty product alarm and low battery signal
- Locking dispenser contains a translucent window for product visibility without product tampering
Specifica
Hybrid Dispenser (Manual/Automatic) | |
Black |