missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS4L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549111
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
INTS4L1 Polyclonal antibody specifically detects INTS4L1 in Human samples. It is validated for Western Blot.Specifica
INTS4L1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
0 | |
INTS4P1 | |
KLH conjugated synthetic peptide between 232-258 amino acids from the Central region of human INTS4L1/INTS4L2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
INTS4P1 | |
integrator complex subunit 4 pseudogene 1; INTS4L1; INTS4P1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
285905 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |