Learn More
Ismatec™ Reglo Digital Miniflex Pumps
Codice prodotto. 16436649
Click to view available options
Portata:
0.004 to 190 mL/min.
0.004 to 365 mL/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 16436649
Marca: Ismatec™ 7801842
Versatile pumps with an intuitive touch-screen interface for accurate low-flow fluid delivery.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5 in. (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen, MasterflexLive™, or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
  • Miniflex pump head accepts continuous tubing for clean, uninterrupted fluid paths
Specifica
Canali 2
Control Type Digital Variable Speed
Tipo di display Touchscreen
Valori nominali armadio IP31
Portata 0.004 to 190 mL/min.
Maximum Pressure Dependent on tubing
Motor Speed 0.1 to 350 rpm
Reversible Yes
Controllo esterno RJ45 Ethernet; USB; 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via DB-25
Include Pump drive, pump head, and power adapter with 6 ft. (1.8 m) line cord
Materiale ABS Plastic and Aluminum
Dimensioni (L x P x A) 22.9 x 14.7 x 15.7 cm
Requisiti elettrici 90 to 260 VAC
Tensione 90/260 VAC