Learn More
Trajan™ Jumbo Syringe
Click to view available options
Volume (sistema metrico):
500 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
Descrizione
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
- Designed for holding and dispensing large volumes of gas
- Heavy duty acrylic barrels
- Easy access to sample for the addition of standards or removal of subsample via secondary port
- Plunger stem can be unscrewed for ease of transportation and storage
- Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifica
Specifica
| Materiale | Acrylic, Acrylic (Barrel) |
| Da utilizzare con (applicazione) | For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling |
| Product Type | Jumbo Syringe |
| Modello | 500MAR-LL-GT |
| Volume (sistema metrico) | 500 mL |