Learn More
K-425 Model SpeedDigestor
Set new standards in terms of digestion speed, flexible use and result reproducibility with the IR SpeedDigester. BUCHI K-425 Model SpeedDigestors provide increased sample throughput for nitrogen determination by Kjeldahl digestion and multiply instrument functionality when needs change.
Marca: Buchi 1154252200
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Fast heat transfer of infrared heaters to samples
- Large time savings due to fast heating and cooling
- Increased sample throughput due to shortened process time
- Accelerated digestion step with continuous addition of H2O2
- One instrument for both Kjeldahl and reflux digestion
- All BUCHI sample tubes to be used (100mL, 300mL, 500mL)
- Sample tubes compliant with ISO 6060 for COD or other reflux digestion (e.g. aqua regia)
- Dedicated suction module for aqueous samples
- Sealed suction module for effective transfer of harmful fumes
- Effective neutralization of fumes by the Scrubber K-415
- Safe operation and extended lifetime of fume hood
Specifica
Speed Digestor | |
220/240 V ±10 V | |
620 mm | |
Feed - Kjeldahl, Chemicals/Agricultural, Beverage, Pharmaceutical / Cosmetics , Food , Environmental Analysis | |
310 mm | |
540 mm |
50/60 Hz | |
1280 W | |
220/240 V ±10 V, 50/60 Hz, 1280 W | |
300mL Sample tubes and suction module with condensate trap | |
1 Ea. |