Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Click to view available options
Capacità (sistema metrico):
250 mL
2000 mL
4 L
Dimensione della confezione:
Pezzo
pezzi 2
Descrizione
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
- Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
- Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
- Graduated with approximate volume scale in durable white ceramic enamel
- Tubing and stoppers sold separately
Specifica
Specifica
| Certificazione/Conformità | ASTM E1406, Type II, Class 2 |
| Diametro (sistema imperiale) esterno | 3.26 in. |
| Capacità (sistema metrico) | 250 mL |
| Materiale | Borosilicate Glass |
| Intervallo di graduazione | 50 to 250 mL |
| Graduazioni | 25 mL |
| Altezza (sistema imperiale) | 6.4 in. |
| Altezza (sistema metrico) | 16.3 cm |
| Diametro (sistema metrico) esterno | 8.3 cm |
| Dimensione chiusura | 6 |