Learn More
Kinematica™ H-Foot Stand ST-F 10/600 for Polytron™ PT 3100 D
An H-foot stand made of anodized aluminum.
Marca: Kinematica™ 11040025
Dettagli aggiuntivi : Peso : 6.99300kg
Descrizione
- Height adjustable
- Swiveling PTFE-coated work table with vessel holder
- 5.5kg in weight
Specifica
Homogenizer Stand | |
Anodized Aluminum | |
H-foot homogenizer stand | |
ST-F 10/600 |