Learn More
KIR2DL1/CD158a Antibody (HP-MA4), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-62191
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
KIR2DL1/CD158a Monoclonal antibody specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry, ImmunoprecipitationSpecifica
KIR2DL1/CD158a | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunoprecipitation | |
HP-MA4 | |
Flow Cytometry 1-4 μg/ml, Immunoprecipitation 1:10-1:500 | |
CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
Human NK cell line LB2. CD158a/g/h | |
0.1 mg | |
Immunology, Innate Immunity, Signal Transduction | |
3802 | |
Store at 4°C. Do not freeze. | |
IgG2b κ |