Learn More
Lenz Laborglasintrumente Kjeldahl Flasks
Codice prodotto. 10420323
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
750 mL
1 L
Unit Size:
pezzi 10
Product Code. 10420323
Brand: Lenz Laborglasintrumente 03052964
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the determination of nitrogen in organic compounds
  • With center neck
  • With conical joint and inscription label
  • Material: DURAN™
Specifications
Capacità (sistema metrico) 750 mL
Materiale Borosilicate Glass (3.3)
Giunto rastremato standard NS 29/32
Diametro (sistema metrico) esterno 115 mm
Quantità 10/Pk.