Learn More
BUCHI KjelMaster™ Automated Distillation Unit
Marca:  BUCHI 113751700
Dettagli aggiuntivi : Peso : 39.83000kg
Codice prodotto. 15376221
  • 52745.00€ / Pezzo
Spedizione stimata: 11-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Order Info
Model
Specifica
Specifica
220 to 240V, 50/60Hz
220 to 240V
431mm
Kjeldahl Distillation Automated System
K-375
458mm
Potentiometric titration: 300mL Sample tube, Pair of glass tongs, pH electrode, 3 x 20mL each Buffer set pH 4 and pH 7, 250mL KCI electrolyte, saturated, Bottle (1L) for titrant solution, (2) Storage tanks (10L) for chemicals, (2) Storage tanks (20L) for water and waste, (4) Capacitive level sensors for tanks with o-rings, Hose connector for waste tank, 6m Hose for chemical feed, 3 Suction tubes to tanks, 1.8m Hose waste drain, 1.5m Hose for cooling water feed, 1.8m Hose for cooling water drain, 2m Connection cable RJ45, Mini gender changer for balance connection, Test gauge sample tube holder, Connection grommet for Chiller, 20 Weighing boats, CD KjelLink PC software (with 60 days test license)
670mm
1Each