missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid 5-ethyl ester, 95%, Thermo Scientific™
Click to view available options
Quantità:
5 g
25 g
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 100 mg/ml.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 100 mg/ml.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Specifica
Materiale o nome chimico | L-Glutamic acid 5-ethyl ester |
Punto di fusione | 179°C (decomposition) |
Colore | White |
Informazioni di solubilità | Soluble in water 100mg/ml. |
IUPAC Name | (2S)-2-amino-5-ethoxy-5-oxopentanoic acid |
Peso formulazione | 175.18 |
Percent Purity | 95% |
Quantità | 25 g |
Forma fisica | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'