Learn More
PTEN/MMAC1 Ab-4 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ PTEN/MMAC1 Ab-4, Mouse Monoclonal Antibody.
Marca: Epredia MS-1601-S
Descrizione
PTEN/MMAC1, a tumor suppressor, has been implicated in a large number of human tumors and is conserved from humans to worms. Characterization of PTEN protein showed that it is a phosphatase that acts on proteins and on 3-phosphorylated phosphoinositides, and can therefore modulate signal-transduction pathways that involve lipid second messengers. Recent results indicate that at least part of its role is to regulate the activity of the serine/threonine kinase AKT/PKB, and thus influence cell survival signaling.Host Species: Mouse
Clone: 17.A
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human PTEN protein
Molecular Weight: 60kDa
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifica
| PTEN/MMAC1 Ab-4 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Cancer and Tumor Biology | |
| Human | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| 17.A | |
| 1:25-1:50 for 30 min at RT | |
| Recombinant human PTEN protein | |
| RUO | |
| Primary | |
| Store vial at 4deg.C. When stored at 2 to 8deg.C, | |
| IgM κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.