Learn More
Labconco™ FilterMate™ Portable Exhauster
Labconco FilterMate™ Portable Exhauster removes hazardous powders, particulates or vapors from exhaust air stream, returning filtered air to environment.
Marca: Labconco™ 3970023
Dettagli aggiuntivi : Peso : 5.00000kg
Includes: 8 ft. of 5 in. I.D. gray flexible hose, two clamps and one 5 in. O.D. Upper Transition Adaptor for connection to Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
Requires : Carbon Filter (Models for use with carbon filter only). Models for use with carbon filter require Organic Vapor Cell, Formaldehyde Impregnated Cell, or Ammonia Impregnated Cell. Combination models can support use of two carbon filters by removing HEPA filter.
Descrizione
An accessory for Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
- Compact, corrosion-resistant housing of glacier white and light gray, dry powder, epoxy-coated steel with rubber feet
- Uses HEPA filter, carbon-based filter or combination of both filters
- Variable speed, non-sparking motorized impeller, adjustable to optimize airflows
- Negative pressure plenum surrounds positive pressure impeller to capture and refilter unfiltered air
- Front-mounted rocker switch controls power to blower
- Rear-mounted auxiliary electrical outlet for connection to airflow monitor
- Suitable for 2 ft., 3 ft. and 4 ft. wide x 22.75 in. high enclosures
- Comes with IEC 3-wire electrical cord with plug
Models for use with HEPA filter
- Include true bag-in bag-out filter disposal system for safely removing and replacing filter without detaching hose
- Filter efficiency: 250 CFM
Filter efficiency: 250 CFM
Models for use with HEPA and Carbon-based filters
Filter efficiency: 200 CFM
Compliance
UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifica
Exhauster | |
CE | |
250 cfm | |
58 cm | |
14.9 in. | |
230 V | |
22.7 kg | |
58 cm |
Ammonia-Amine, Formaldehyde, HEPA, Organic Vapor | |
14.9 x 23 x 22.7 in. (38 x 58 x 58 cm) | |
22.7 in. | |
50 Hz | |
38 cm | |
50 lb. | |
23 in. |