Learn More
Labconco™ Protector Glove Box Accessory Kits
Kits provided added functionality options for Protector glove boxes
1000.00€
Codice del prodotto | Marca | Include | Product Type | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Include | Product Type | Prezzo | Quantità e disponibilità | ||||
16649183
|
Labconco™
|
HEPA filter, hose barb connectors and 10 ft. of 0.75 in. ID neoprene tubing for attachment to the pressure relief bubbler exhaust | Pressure Relief Kit |
1000.00€
Pezzo |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Add-A-Valve Kits include two manual valves, tubes, nuts and instructions for adding valves to the glove box for evacuating with vacuum, filling with inert gas, connecting to a drying train, or connecting to filtration components. Manual valves mount in predrilled holes behind the label located below the control panel. For use with Protector Combination and Protector Filtered Glove Boxes.Specifica
Protector Glove Boxes |