Learn More
Labconco™ Kjeldahl Replacement Parts
Replacement parts for Labconco Kjeldahl Digestion and Distillation Apparatus.
Marca:  Labconco™ 2128800
Dettagli aggiuntivi : Peso : 0.20000kg
Codice prodotto. 12753997
  • 243.00€ / pezzi 6
Spedizione stimata: 15-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Tubes feature long necks, round perforated bottoms and thick walls to help eliminate pressure fluctuations in the distillation phase. One-piece borosilicate glass. Each tube may be label with 21288-01. Six per package. For use with Labconco Hooded Combination Kjeldahl Units, Kjeldahl Digestion Units, Kjeldahl Distillation Units, Open Combination Kjeldahl Units and RapidStill II.
Specifica
Specifica
Delivery Tube
1.4 kg
RapidStill II Unit
Borosilicate Glass
3 lb.