Learn More
Labconco™ SteamScrubber™ Culture Tube Inserts
Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.
Descrizione
Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.
Type 304 stainless steel basket-style Inserts fit in standard open racks and spindle racks in SteamScrubber and FlaskScrubber Glassware Washers. Holds 144 each 15 to 18 mm OD tubes. Grid size is 23.8 mm x 23.8 mm. For use in lower rack only.Specifica
| Each | |
| Stainless Steel (304) |