Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Marca: Thermo Scientific™ SS00158-I
Dettagli aggiuntivi : Peso : 5.60000kg
Descrizione
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site). It is available in 1, 5, 10 and 20L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: TPE 374, length: 30.5cm (12 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC insert + cap, pinch clamp. Tubing: TPE 374, length: 91.
Specifica
Store at room temperature. | |
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
TPE | |
Two 4-mil. poly bags | |
Labtainer™ | |
15/Cs. | |
Irradiated | |
32 cm | |
14.7 cm |
1 L | |
Polyethylene | |
ASI™ 26/77 | |
Capsule filter and MPC insert + cap | |
Protective material around rough areas | |
Room Temperature | |
32 x 14.7 cm (12.6 x 5.8 in.) | |
5.8 in. |