Learn More
Brady™ Laminated polyester: ISO Safety Sign - Direction arrow 45°, Top Right Corner, Red
Sign compliant to ISO 7010.
Marca: Brady™ 830822
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Laminated polyester
- For indoor and outdoor use
Specifica
ISO 7010 | |
Polyester | |
Self-adhesive | |
148 mm | |
148 mm | |
Square | |
White on Red |
Gloss | |
Permanent Acrylic | |
Indoor and Outdoor | |
0.099 mm | |
-40°C to 110°C | |
Global |