Learn More
Euro Protection™ Latex Gloves, Powder-Free
Codice prodotto. 15322286
Click to view available options
Size:
6
8
10
12
Euro Protection™ Latex Gloves, Powder-Free are comfortable, flexible, and suitable for food contact.
  • Liquidproof
  • Good dexterity
  • Comfort and flexibility
  • Suitable for food contact
Specifica
Certifications/Compliance (Europe) EN374-1:2016K(3/6)TYPE C EN374-5:2016 EN420
Monouso Single-use
Materiale Latex
Powdered Powder Free
Size 12
Sterilità Non-sterile
Spessore (sistema metrico) 0.125 mm
Da utilizzare con (applicazione) Object handling, food industry, paint, solvent, liquid (non-toxic), assembly, cleaning, hospital environments (laboratory)
Quantità 100/Pk