Learn More
Hach Lange™ Sensor Replacement Kits for LDO101 Probes
Robust and simple to use, this LDO dissolved oxygen (DO) sensor from Hach has made measuring DO in many samples much easier.
Marca: Hach Lange™ 5811200
Dettagli aggiuntivi : Peso : 0.08500kg
Descrizione
- LDO Sensor Replacement Kit
- For use with Hach LDO101 Standard and Rugged Dissolved Oxygen Probes
- The LDO dissolved oxygen sensor from Hach has made measuring DO in many samples much easier
- Robust and simple to use
Specifica
Dissolved Oxygen Probe | |
LDO Sensor Replacement Kit |