Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Basic controller MC 750
Offers continuous dimming and over-temperature protection of the connected LED light head. The VisiLED Controllers are the core of the VisiLED system to control the LED light heads.
Marca: SCHOTT AG Lighting and Imaging™ 400010
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Basic controller with power on/off switch
- Knob turns left or right for dimming
- Dimmable from 0 to 100%
- For use with EasyLED and VisiLED Series LED Illumninators
Specifica
Controller for VisiLED Light Heads | |
Stereo Microscopy Illumination | |
VisiLED Light Heads |