Learn More
Thermo Scientific Chemicals Low Form Inconel Crucible;Cap (ml), 500;Top Dia (mm), 102;Bottom Dia (mm), 89;Depth (mm), 66
Click to view available options
Capacità (sistema metrico):
15 mL
25 mL
55 mL
75 mL
100 mL
250 mL
500 mL
Diametro (sistema metrico) esterno:
33mm
45mm
46mm
47mm
51mm
59mm
82mm
102mm
Altezza (sistema metrico):
23mm
35mm
43mm
46mm
60mm
66mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Descrizione | Low Form |
| Materiale | Inconel |
| Capacità (sistema metrico) | 250 mL |
| Diametro (sistema metrico) esterno | 82mm |
| Altezza (sistema metrico) | 60mm |
| Product Type | Inconel Crucible |
| Diametro (sistema metrico) base | 66mm |