Learn More
Low UV Standard for ICP, 3 Components, Honeywell Fluka™
Marca: Honeywell Fluka E2125-100ML
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifica
Low UV Standard for ICP | |
1.06 g/cm3 | |
Standards for ICP and ICP-MS |
100 mL | |
<2 |