Learn More
BUCHI Lyovapor L-200 Shelves 6
Codice prodotto. 17274724 Sfoglia Tutto Buchi Prodotti
Click to view available options
Include:
Advanced vacuum control, No Sample temperature sensor
Advanced vacuum control, No Sample temperature sensor, Trolley
Pompe da vuoto:
Edwards nXDS6iC
No Vacuum Pump
Pfeiffer Duo 6
Dimensione della confezione:
Pezzo
Codice prodotto. 17274724
Marca: BUCHI 11L200IN1612001101
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Offers high-quality Freeze Drying (-55°C, 6 kg) with a high level of automation. Infinite-Control™ includes easy method creation, method running, data logging, chart recording in real-time and interruption if needed from anywhere at anytime.
  • Compact Lyovapor™ L-200
  • Offers high-quality Freeze Drying (-55 °C, 6 kg) with a high level of automation.
  • Infinite-Control™ includes easy method creation, method running, data logging, chart recording in real-time and interruption if needed from anywhere at anytime.
Specifica
Tipo Freeze Dryer
Capacità (sistema metrico) 6 kg
N. di ripiani 6
Temperature Range (Metric) -55°C to +30°C
Materiale Stainless Steel
Software Included No
Pompe da vuoto Pfeiffer Duo 6
Include Advanced vacuum control, No Sample temperature sensor, Trolley
Altezza (sistema imperiale) 20.0787 in.
Lunghezza (sistema imperiale) 23.1496 in.
Tipo di spina All except North America
Tensione 220/240 V