Learn More
Cytiva (formerly Pall Lab) Magnetic Filters Funnel, Non-sterile
Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.
Marca: Cytiva (formerly Pall Lab) 4242
Dettagli aggiuntivi : Peso : 0.28500kg
Descrizione
- Not approved for use in medical, clinical, surgical or other patient applications
Specifica
300 mL | |
Polyphenylsulfone | |
EFA: 9.6 cm2 | |
47 mm |