missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Mastercycler Nexus X2e, EU-Plug

PCR cycler with the capacity to run two simultaneous and independent protocols.

Marca:  Eppendorf™ 6339000016


Dettagli aggiuntivi : Peso : 11.00000kg

Codice prodotto. 15548831

  • 7575.00€ / Pezzo
In stock
per visualizzare lo stock disponibile

Descrizione

Descrizione

  • Large block for large assays - small block for small assays
  • Intuitive graphic programming
  • Universal block for strips, 0.1 mL, 0.2 mL, and 0.5 mL PCR tubes
  • Small footprint
  • Optional gradient for PCR optimization
  • E-mail notification
  • flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
Specifica

Specifica

37 to 110°C (Lid), 4 to 99°C (Block)
25 × 41.2 × 33 cm
230 V, 50/60 Hz
CAN_in/CAN_out, Ethernet
Yes
EU Plug
64/32 × 0.2 mL PCR Tubes or 24/10 × 0.5 mL PCR Tube Positions
Standard PCR, Optimizing a PCR
>700
11 kg
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali