Learn More
WTW™ Measuring Vessel
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Marca: WTW™ 209510
Dettagli aggiuntivi : Peso : 0.70000kg
Specifica
1000 mL | |
Respirometry |