Learn More
MES lysis buffer with NP-40 (2X), Thermo Scientific™
Click to view available options
Quantità:
125 mL
250 mL
Dimensione della confezione:
125mL
250mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | MES lysis buffer with NP-40 |
| Concentrazione | 2X |
| Forma fisica | Liquid |
| Quantità | 125 mL |