missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl oleate, tech. C18 71-90%, C18:1 >65% of C18, Thermo Scientific Chemicals
Click to view available options
Quantità:
250 mL
1000 mL
Descrizione
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifica
Materiale o nome chimico | Methyl oleate |
Densità | 0.876 |
Punti di ebollizione | 168°C to 170°C (2 mmHg) |
Punto d'infiammabilità | 163°C (325°F) |
Odore | Weak, Characteristic |
Note percentuale saggio | C18 71 to 90%, C18:1 >65% of C18 |
Formula lineare | CH3(CH2)7CH=CH(CH2)7CO2CH3 |
Indice di rifrazione | 1.452 |
Quantità | 250 mL |
Beilstein | 1727037 |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'