Learn More
Emma Safety Footwear Metric Safety Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifica
Specifica
| Certificazione/Conformità | EN ISO 20345:2011, EN 12568:2010 |
| Tipo di chiusura | Elastic |
| Colore | White |
| Progettazione | Metric |
| Materiale rivestimento interno | Hydro-Tec™ Sanitized Silver |
| Materiale | Microfiber |
| Size | 37 |
| Materiale dello zoccolo | Steel |