Learn More
Metro™ Gowning Benches
Sturdy and comfortable gowning fixtures
Marca:  Metro™ GB1648S
Dettagli aggiuntivi : Peso : 19.96000kg
Codice prodotto. 15996911
  • 1440.00€ / Pezzo
Spedizione stimata: 11-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • 18 in. high (45.7cm) bench with heavy-duty, solid top made of 14 gauge, Type 304 stainless-steel
  • Adjustable stainless-steel foot plates provide for floor mounting
  • Nonconductive but can be made conductive with the addition of aluminum split sleeves (Cat. No. 17-987-44)
Specifica
Specifica
16 in.
55 lb.
40.64 cm
121.9 cm
Stainless Steel
25 kg
45.72 cm
18 in.
Silver